Check Order Status
PBS, Phosphate Buffered Saline, 1X Solution, pH 7.4, Fisher BioReagents™
Actual product may vary
Fisher Chemical
PBS, Phosphate Buffered Saline, 1X Solution, pH 7.4, Fisher BioReagents™
Have Questions?
Catalog Number
Quantity
FGCBP2438-4
4L
Catalog number
FGCBP2438-4
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4L
PBS, Phosphate Buffered Saline, 1X Solution, pH 7.4
A standard phosphate buffer used in many biomolecular procedures.
Specifications
Packaging Type
Poly Cube
Melting Point
0°C
Quantity
4L
pH
7.3 to 7.5
Form
Liquid
Unit Size
EA
Contents & Storage
Room temperature