Check Order Status
TBE Buffer Mix (Dry Powder Mix of Tris/Boric Acid/EDTA/Electrophoresis), Fisher BioReagents
Element Search
Structure Search
TBE Buffer Mix (Dry Powder Mix of Tris/Boric Acid/EDTA/Electrophoresis), Fisher BioReagents
Have Questions?
Catalog Number
Quantity
FGCBP1396-86
86.1 g
Catalog number
FGCBP1396-86
Price (MYR)
-
Quantity:
86.1 g
Request bulk or custom format
Chemical Identifiers
CAS
77-86-1, 6381-92-6, 10043-35-3
Specifications
Chemical Name or Material
TBE Buffer Mix
CAS
77-86-1
ChemAlert Storage Symbol
Gray
Color
White
CAS
6381-92-6
View more
Electrophoresis buffer.