Check Order Status
N-Butylamine, Extra Pure, SLR, Fisher Chemical
Element Search
Structure Search
N-Butylamine, Extra Pure, SLR, Fisher Chemical
Have Questions?
Catalog number
FGCB5260PB07
also known as FGCB/5260/PB07
Price (MYR)
-
Request bulk or custom format
Specifications
CAS
109-73-9
Melting Point
-49°C
Color
Colorless
pH
12.6
Boiling Point
78°C
View more