Check Order Status
Sec-Butyl Alcohol, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Sec-Butyl Alcohol, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FGCB520015
also known as FGCB/5200/15
1 L
Amber glass bottle
Catalog number
FGCB520015
also known as FGCB/5200/15
Price (MYR)
-
Quantity:
1 L
Packaging:
Amber glass bottle
Request bulk or custom format
Specifications
CAS
78-92-2
Melting Point
-115°C
Color
Colorless
Boiling Point
99°C
Packaging
Amber glass bottle
View more