Check Order Status
1-Butane Sulfonic Acid, Sodium Salt, for HPLC, Certified, Fisher Chemical
Element Search
Structure Search
1-Butane Sulfonic Acid, Sodium Salt, for HPLC, Certified, Fisher Chemical
Have Questions?
Catalog number
FGCB479146
also known as FGCB/4791/46
Price (MYR)
655.00
BOT
Add to cart
Request bulk or custom format
Price (MYR)
655.00
BOT
Add to cart
Specifications
Melting Point
300°C
CAS
2386-54-1
Color
White
Physical Form
Powder/Solid
Quantity
25 g
View more