Check Order Status
Bromine, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Bromine, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCB4000PB05
also known as FGCB/4000/PB05
100 mL
Catalog number
FGCB4000PB05
also known as FGCB/4000/PB05
Price (MYR)
701.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
701.00
EA
Add to cart
Specifications
Boiling Point
58.7°C
Melting Point
-7.2°C
CAS
7726-95-6
Color
Brown-Red
Physical Form
Liquid
View more