Check Order Status
Benzyl Alcohol, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
Element Search
Structure Search
Benzyl Alcohol, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FGCB2550PB17
also known as FGCB/2550/PB17
2.5 L
Plastic Coated Glass bottle
Catalog number
FGCB2550PB17
also known as FGCB/2550/PB17
Price (MYR)
-
Quantity:
2.5 L
Packaging:
Plastic Coated Glass bottle
Request bulk or custom format
Specifications
CAS
100-51-6
Melting Point
-15°C
Color
Yellow
Boiling Point
205°C
Packaging
Plastic Coated Glass bottle
View more