Check Order Status
Benzoyl Chloride, Certified AR for Analysis, Fisher Chemical
Element Search
Structure Search
Benzoyl Chloride, Certified AR for Analysis, Fisher Chemical
Have Questions?
Catalog number
FGCB2350PB08
also known as FGCB/2350/PB08
Price (MYR)
-
Request bulk or custom format
Specifications
CAS
98-88-4
Melting Point
-1°C
Color
Colorless
pH
2
Boiling Point
198°C
View more