Check Order Status
Barium Hydroxide Octahydrate, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Barium Hydroxide Octahydrate, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCB065060
also known as FGCB/0650/60
1 kg
Catalog number
FGCB065060
also known as FGCB/0650/60
Price (MYR)
-
Quantity:
1 kg
Request bulk or custom format
Specifications
Melting Point
78°C
CAS
12230-71-6
Color
White
pH
12.5
Physical Form
Solid
View more