Check Order Status
Barium Carbonate, 99+%, Extra Pure, SLR, Fisher Chemical
Element Search
Structure Search
Barium Carbonate, 99+%, Extra Pure, SLR, Fisher Chemical
Have Questions?
Catalog number
FGCB035053
also known as FGCB/0350/53
Price (MYR)
-
Request bulk or custom format
Specifications
Boiling Point
1450°C
Melting Point
811°C
CAS
513-77-9
Color
White
Physical Form
Powder/Solid
View more