CD4 Recombinant Rabbit Monoclonal Antibody (BL-155-1C11)
CD4 Recombinant Rabbit Monoclonal Antibody (BL-155-1C11)
Actual product may vary
CD4 Recombinant Rabbit Monoclonal Antibody (BL-155-1C11)
CD4 Recombinant Rabbit Monoclonal Antibody (BL-155-1C11)
CD4 Recombinant Rabbit Monoclonal Antibody (BL-155-1C11)
CD4 Recombinant Rabbit Monoclonal Antibody (BL-155-1C11)
CD4 Recombinant Rabbit Monoclonal Antibody (BL-155-1C11)
Rabbit Recombinant Monoclonal Antibody
Have Questions?
Catalog NumberQuantity
FGCA700015
also known as FGCA/7000/15
100 μL
Catalog number FGCA700015
also known as FGCA/7000/15
Price (MYR)
417.00
EA
Add to cart
Quantity:
100 μL
Price (MYR)
417.00
EA
Add to cart
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections.
The CD4 antigen is involved in the recognition of MHC class II molecules and is a co-receptor for HIV. CD4 is primarily expressed in a subset of T-lymphocytes, also referred to as T helper cells, but may also be expressed by other cells in the immune system, such as monocytes, macrophages, and dendritic cells. At the tissue level, CD4 expression may be detected in thymus, lymph nodes, tonsils, and spleen, and also in specific regions of the brain, gut, and other non-lymphoid tissues. CD4 functions to initiate or augment the early phase of T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase, Lck. It may also function as an important mediator of direct neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcripts have been identified in this gene [RefSeq, July 2017].
Specifications
AntigenCD4
ApplicationsFlow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
ClassificationRecombinant Monoclonal
CloneBL-155-1C11
Concentration1 mg/mL
ConjugateUnconjugated
FormulationBBS with 0.09% sodium azide; pH 8.2
GeneCD4
Gene Accession No.P01730
Gene AliasActivation B7-1 antigen; B7; B7.1; B7-1; BB1; B-lymphocyte activation antigen B7; CD28LG; CD28LG1; CD4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; Cd4 molecule; CD4 precursor; CD4 receptor; CD4, allele 1; cd4a; CD4mut; CD80; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); CD80 molecule; cell surface glycoprotein CD4; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; fCD4; L3T4; LAB7; Leu-3; Ly-4; lymphocyte antigen CD4; lymphocyte antigen CD4 precursor; membrane protein; p55; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); T-lymphocyte activation antigen CD80; W3/25; W3/25 antigen
Gene SymbolsCD4
Host SpeciesRabbit
ImmunogenRecombinant protein representing the extracellular domain of human CD4 (residues 26-397).
Purification Methodpurified
Quantity100 μL
Regulatory StatusRUO
Primary or SecondaryPrimary
Gene ID (Entrez)920
Target SpeciesHuman
Product TypeAntibody
FormLiquid
IsotypeIgG
Unit SizeEA
Contents & Storage
4°C