Check Order Status
Ammonium Nickel(II) Sulfate Hexahydrate, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Ammonium Nickel(II) Sulfate Hexahydrate, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCA580050
also known as FGCA/5800/50
250 g
Catalog number
FGCA580050
also known as FGCA/5800/50
Price (MYR)
-
Quantity:
250 g
Request bulk or custom format
Specifications
CAS
15699-18-0
Color
Blue to Green
pH
4.6
Physical Form
Solid
Quantity
250 g
View more