Check Order Status
Di-Ammonium Hydrogen Orthophosphate, Certified AR for analysis, Fisher Chemical™
Element Search
Structure Search
Di-Ammonium Hydrogen Orthophosphate, Certified AR for analysis, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCA536053
also known as FGCA/5360/53
500 g
Catalog number
FGCA536053
also known as FGCA/5360/53
Price (MYR)
-
Quantity:
500 g
Request bulk or custom format
Specifications
CAS
7783-28-0
Color
White
pH
7.9 to 8.3
Physical Form
Crystalline Solid
Quantity
500 g
View more
Di-Ammonium Hydrogen Orthophosphate, Certified AR for analysis