Check Order Status
Ammonium Hydrogen Difluoride, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Ammonium Hydrogen Difluoride, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 kg
500 g
Catalog number
FGCA528062
also known as FGCA/5280/62
Price (MYR)
648.00
EA
Add to cart
Quantity:
2.5 kg
Request bulk or custom format
Price (MYR)
648.00
EA
Add to cart
Specifications
Boiling Point
230°C
Melting Point
125°C
CAS
1341-49-7
Color
White
pH
3.5
View more