Check Order Status
Ammonium Iron(II) Sulfate Hexahydrate, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Ammonium Iron(II) Sulfate Hexahydrate, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCA484063
also known as FGCA/4840/63
3 kg
Catalog number
FGCA484063
also known as FGCA/4840/63
Price (MYR)
-
Quantity:
3 kg
Request bulk or custom format
Specifications
Melting Point
100°C
CAS
7783-85-9
Color
Green
pH
3,5
Physical Form
Solid
View more