Check Order Status
Hydrofluoric Acid 47-51%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Hydrofluoric Acid 47-51%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
500 mL
250 mL
Catalog number
FGCA463-500
Price (MYR)
5,180.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
5,180.00
EA
Add to cart
Specifications
pH
1
Color
Colorless
CAS
7664-39-3
Melting Point
-35°C
Boiling Point
105°C
View more