Check Order Status
Ammonium Chloride, Certified AR, for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical
Element Search
Structure Search
Ammonium Chloride, Certified AR, for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCA392060
also known as FGCA/3920/60
1 kg
Plastic powder jar
Catalog number
FGCA392060
also known as FGCA/3920/60
Price (MYR)
513.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Plastic powder jar
Request bulk or custom format
Price (MYR)
513.00
EA
Add to cart
Specifications
Melting Point
340°C
CAS
12125-02-9
Color
White
pH
5
Physical Form
Solid
View more