Check Order Status
Ammonia Solution, 35%, for HPLC, Fisher Chemical™
Element Search
Structure Search
Ammonia Solution, 35%, for HPLC, Fisher Chemical™
H5NO, CAS Number-1336-21-6, Colorless, 35.05g/mol, 11418, Plastic Coated Glass bottle, 12, Liquid | CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Catalog number
FGCA3295PB05
also known as FGCA/3295/PB05
Price (MYR)
268.00
Price (MYR)
268.00
EA
Add to cart
Specifications
pH
12
Color
Colorless
CAS
1336-21-6
Molecular Formula
H5NO
MDL Number
MFCD00066650
View more
H5NO, CAS Number-1336-21-6, Colorless, 35.05g/mol, 11418, Plastic Coated Glass bottle, 12, Liquid