Check Order Status
Ammonia Solution, 35%, Extra Pure, SLR, d=0.88, Fisher Chemical™
Element Search
Structure Search
Ammonia Solution, 35%, Extra Pure, SLR, d=0.88, Fisher Chemical™
H5NO, CAS Number-1336-21-6, 2.5L, Colorless, 35.05g/mol, 11418, Plastic Coated Glass bottle, 12, Liquid
Have Questions?
Catalog Number
Quantity
FGCA3240PB15
also known as FGCA/3240/PB15
1 L
Catalog number
FGCA3240PB15
also known as FGCA/3240/PB15
Price (MYR)
310.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
310.00
EA
Add to cart
Specifications
pH
12
Color
Colorless
CAS
1336-21-6
Molecular Formula
H
5
NO
MDL Number
11418
View more
H5NO, CAS Number-1336-21-6, 2.5L, Colorless, 35.05g/mol, 11418, Plastic Coated Glass bottle, 12, Liquid