Check Order Status
3-AminomethylAlizarin-N,N-Diacetic Acid, Pure, Indicator Grade, Fisher Chemical
Element Search
Structure Search
3-AminomethylAlizarin-N,N-Diacetic Acid, Pure, Indicator Grade, Fisher Chemical
Have Questions?
Catalog number
FGCA293043
also known as FGCA/2930/43
Price (MYR)
-
Request bulk or custom format
Specifications
CAS
3952-78-1
Melting Point
190°C
Color
Orange to Yellow
Packaging
Glass Jar
Molecular Formula
C
19
H
15
NO
8
View more