Check Order Status
Aluminum Chloride Hydrate, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical
Element Search
Structure Search
Aluminum Chloride Hydrate, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical
Have Questions?
Catalog number
FGCA196053
also known as FGCA/1960/53
Price (MYR)
433.00
Price (MYR)
433.00
EA
Add to cart
Specifications
CAS
7784-13-6
Color
White
pH
2.5 to 3.5
Physical Form
Solid
Quantity
500 g
View more
500g, Plastic powder jar