Check Order Status
Aluminum Ammonium Sulfate, Extra Pure, SLR, Fisher Chemical
Element Search
Structure Search
Aluminum Ammonium Sulfate, Extra Pure, SLR, Fisher Chemical
Have Questions?
Catalog number
FGCA184060
also known as FGCA/1840/60
Price (MYR)
-
Request bulk or custom format
Specifications
Boiling Point
200°C
Melting Point
93.5°C
CAS
7784-26-1
Color
White
pH
4.6
View more