Check Order Status
Ammonium Formate (Optima LC/MS), Fisher Chemical
Element Search
Structure Search
Fisher Chemical
Ammonium Formate (Optima LC/MS), Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCA115-50
50 g
Glass Bottle
Catalog number
FGCA115-50
Price (MYR)
432.00
EA
Add to cart
Quantity:
50 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
432.00
EA
Add to cart
Specifications
Chemical Name or Material
Ammonium Formate
Melting Point
115°C
CAS
540-69-2
Color
White
pH
6.0 to 7.0
View more