Check Order Status
Adipoyl Chloride, Pure, Fisher Chemical
Actual product may vary
Adipoyl Chloride, Pure, Fisher Chemical
Have Questions?
Catalog number
FGCA102099
also known as FGCA/1020/99
Price (MYR)
865.00
EA
Add to cart
Price (MYR)
865.00
EA
Add to cart
Specifications
CAS
111-50-2
Color
Brown
Boiling Point
105°C
Packaging
Ampoule
Molecular Formula
C
6
H
8
Cl
2
O
2
MDL Number
759
Quantity
5 mL
Formula Weight
183.03g/mol
Physical Form
Liquid