Check Order Status
Acetone, Technical, Fisher Chemical™
Element Search
Structure Search
Acetone, Technical, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FGCA052017
also known as FGCA/0520/17
2.5 L
Amber glass bottle
Catalog number
FGCA052017
also known as FGCA/0520/17
Price (MYR)
291.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Amber glass bottle
Request bulk or custom format
Price (MYR)
291.00
EA
Add to cart
Chemical Identifiers
CAS
67-64-1
Specifications
Packaging
Amber glass bottle
CAS
67-64-1
Vapor Pressure
247mbar at 20°C
Viscosity
0.32 mPaS at 20°C
Vapor Density
2
View more