Filling Solution: Saturated KCl, 29.8%
Product Image
Filling Solution: Saturated KCl, 29.8%
For calomel single-junction electrodes; outer chamber of Ag/AgCl double-junction electrodes; Fluoride, Sodium Combination ISEs, accupHast and accuTupH electrode outer chambers.
Have Questions?
Catalog NumberBoiling Point
FCSSP138-500100°C
Catalog number FCSSP138-500
Price (MYR)
-
Boiling Point:
100°C
  • Saturated KCl electrolyte
  • For all calomel single-junction electrodes
  • For outer chamber of Ag/AgCl double-junction electrodes
  • For Fluoride Combination ISEs 13-620-528 and 13-620-529
  • For Sodium Combination ISEs 13-620-502, 13-620-503
  • For outer chanber of accupHast, accupHast R, accuTupH, accuTupH plus double-junction electrodes
  • In 500mL HDPE plastic bottle
Specifications
Boiling Point100°C
Capacity (English)16.9 oz.
Capacity (Metric)500 mL
ColorColorless
DescriptionSaturated at 20°C
For Use With (Equipment)Calomel single-junction Electrode, outer chamber of Ag/AgCl double-junction Electrode, Fluoride, Sodium combination ISEs, AccupHast™ and AccuTupH™ Electrode outer chamber
Filling Solution TypeSaturated KCl
Melting Point-10°C
PackagingHDPE Bottle
Water SolubleSoluble
Unit SizeEA