Check Order Status
Buffer Solution, pH 10.00, Color-Coded Blue (Certified), Fisher Chemical™
Element Search
Structure Search
Buffer Solution, pH 10.00, Color-Coded Blue (Certified), Fisher Chemical™
Have Questions?
Change view
Quantity:
500 mL
Packaging:
Poly Bottle
Catalog number
FCSSB115-500
Price (MYR)
-
Quantity:
500 mL
Packaging:
Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
6381-92-6, 584-08-7, 1332-77-0, 1310-58-3
Specifications
Absorbance
Pass Test
Chemical Name or Material
Buffer Solution, pH 10.00, Color-Coded Blue
ChemAlert Storage Symbol
Gray
Melting Point
0°C
Boiling Point
100°C
View more