Check Order Status
Buffer Solution, pH 4.00, Color-Coded Red (Certified), Fisher Chemical™
Element Search
Structure Search
Buffer Solution, pH 4.00, Color-Coded Red (Certified), Fisher Chemical™
pH/temperature conversions given on label
Have Questions?
Change view
Quantity:
500 mL
Packaging:
Poly Bottle
Catalog number
FCSSB101-500
Price (MYR)
-
Quantity:
500 mL
Packaging:
Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
50-00-0, 67-56-1, 877-24-7, 7732-18-5, 16423-68-0
Specifications
Absorbance
Pass Test
Chemical Name or Material
Buffer Solution, pH 4.00, Color-Coded Red
CAS
50-00-0
ChemAlert Storage Symbol
Gray
Melting Point
0°C
View more