Check Order Status
Water, USP/EP Purified, Ricca Chemical
Element Search
Structure Search
Water, USP/EP Purified, Ricca Chemical
Have Questions?
Change view
Quantity:
4 L
10 L
Packaging:
Cubitainer
Catalog number
FCSR91900004F
Price (MYR)
-
Quantity:
4 L
Packaging:
Cubitainer
Request bulk or custom format
Specifications
Chemical Name or Material
Water
Shelf Life
12 Months
pH
5.0 to 7.0
CAS
7732-18-5
Color
Clear, Colorless
View more
Water, USP/EP Purified