Check Order Status
Silver Nitrate, For Chloride, (1mL = 0.5mg Cl
-
), Certified, 0.0141N ±0.0002N (0.0141M), LabChem™
Element Search
Structure Search
Silver Nitrate, For Chloride, (1mL = 0.5mg Cl
-
), Certified, 0.0141N ±0.0002N (0.0141M), LabChem™
Have Questions?
Catalog Number
Quantity
FCSLC226001
500 mL
Catalog number
FCSLC226001
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or Material
Silver Nitrate
Color
Colorless
Name Note
For Chloride, (1mL = 0.5mg Cl
-
)
CAS
7761-88-8
CAS Max %
99.76
View more