Carbon Standard, (Potassium Hydrogen Phthalate), Certified, 1000ppm ±10ppm Organic (1mL = 1mg C) , LabChem™
Carbon Standard, (Potassium Hydrogen Phthalate), Certified, 1000ppm ±10ppm Organic (1mL = 1mg C) , LabChem™
Carbon Standard, (Potassium Hydrogen Phthalate), Certified, 1000ppm ±10ppm Organic (1mL = 1mg C) , LabChem™
Carbon Standard, (Potassium Hydrogen Phthalate), Certified, 1000ppm ±10ppm Organic (1mL = 1mg C) , LabChem™
Carbon Standard, (Potassium Hydrogen Phthalate), Certified, 1000ppm ±10ppm Organic (1mL = 1mg C) , LabChem™
Carbon Standard, (Potassium Hydrogen Phthalate), Certified, 1000ppm ±10ppm Organic (1mL = 1mg C) , LabChem™
Have Questions?
Catalog NumberQuantityPackaging
FCSLC129107125 mLPoly Bottle
Catalog number FCSLC129107
Price (MYR)
-
Quantity:
125 mL
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCarbon Standard
Name Note1000ppm, Organic (1mL = 1mg C) (Potassium Hydrogen Phthalate)
PackagingPoly Bottle
CAS877-24-7
CAS Max %99.49
View more