Check Order Status
Phosphoric Acid, 25% (v/v), Ricca Chemical
Element Search
Structure Search
Phosphoric Acid, 25% (v/v), Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FCS585132
1 L
Natural Poly Bottle
Catalog number
FCS585132
Price (MYR)
-
Quantity:
1 L
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Phosphoric Acid
Color
Colorless
CAS
7732-18-5
CAS Min %
74.0
Concentration
25% (v/v)
View more