Check Order Status
Chromatography-Column
Actual product may vary
Chromatography-Column
Chromatography-Column with fused filter disc.
Have Questions?
Catalog Number
Column Capacity
Pore Size
FCS3110-4
20 mL
4
Catalog number
FCS3110-4
Price (MYR)
-
Column Capacity:
20 mL
Pore Size:
4
Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Quantity
5/Pk.
Pore Size
4
Column Capacity
20 mL
Material
Borosilicate Glass 3.3
Includes
Fused Filter Disc