Check Order Status
Ferric Chloride CS, USP/EP Yellow Primary Solution, Ricca Chemical
Element Search
Structure Search
Ferric Chloride CS, USP/EP Yellow Primary Solution, Ricca Chemical
1 mL = 45.0 mg FeCl
3
.
6H
2
O
Have Questions?
Catalog Number
Quantity
Packaging
FCS3110-16
500 mL
Amber Glass Bottle
Catalog number
FCS3110-16
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Amber Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Ferric Chloride
Color
Yellow
Name Note
Yellow Primary Solution, Colorimetric Solution
CAS
10025-77-1
CAS
7647-01-0
View more