Check Order Status
Cupric Sulfate CS, USP/EP Blue Primary Solution, Ricca Chemical
Element Search
Structure Search
Cupric Sulfate CS, USP/EP Blue Primary Solution, Ricca Chemical
1 mL = 62.4 mg CuSO
4
.
5H
2
O
Have Questions?
Catalog Number
Quantity
Packaging
FCS2495-4
120 mL
Amber Glass Bottle
Catalog number
FCS2495-4
Price (MYR)
-
Quantity:
120 mL
Packaging:
Amber Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Cupric Sulfate
Color
Blue
Name Note
Blue Primary Solution
CAS
7758-99-8
CAS
7647-01-0
View more