Check Order Status
Color Standard, APHA/Hazen (Platinum-Cobalt), Color = 50, Ricca Chemical
Element Search
Structure Search
Color Standard, APHA/Hazen (Platinum-Cobalt), Color = 50, Ricca Chemical
Have Questions?
Catalog Number
Quantity
FCS2230050-16
500 mL
Catalog number
FCS2230050-16
Price (MYR)
-
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS
7647-01-0, 16921-30-5, 7791-13-1, 7647-01-0, 7732-18-5
Specifications
Chemical Name or Material
Color Standard
Name Note
Color = 50
Packaging
Amber Glass Bottle
CAS Min %
97.0
CAS Max %
100.0
View more