CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers
CO<sub>2</sub> and LN<sub>2</sub> Backup Systems for Ultra-Low Temperature Freezers
Actual product may vary
CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Type
FCO456LEVForma FDE, Revco RDE, HERAfreeze HDE, Thermo Scientific TDE Series (400 box capacity and larger)CO2 Backup
1960446TSE Series, Forma 7000 Series, HFU B Series, HFC Series, Revco CxF Series, TSC SeriesCO2 Backup, Field Installed
FCO300VForma FDE30086, Revco RDE30086, HERAfreeze HDE30086, TDE30086CO2 Backup, Field Installed
FCO4567LEThermo Scientific TSX SeriesCO2 Backup, Field Installed
332353G06TDE Series Chest FreezersCO2 Backup, Factory Installed
Catalog number FCO456LEV
Price (MYR)
26,725.00
EA
Add to cart
For Use With (Equipment):
Forma FDE, Revco RDE, HERAfreeze HDE, Thermo Scientific TDE Series (400 box capacity and larger)
Type:
CO2 Backup
Price (MYR)
26,725.00
EA
Add to cart
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers, for use with Thermo Scientific™ TSX, Forma™, HERAfreeze™, Revco™ and TS-Series freezers.
Features:
  • LN2 backup system maintains temperature down to -80°C with liquid nitrogen
  • CO2 backup system maintains temperature down to -67°C with CO2
Ordering Information:
Available as field- or factory-installed. Models for 300 box capacity models are standalone. Field-installed units must be installed by qualified professionals.
Specifications
TypeCO2 Backup
Temperature Range-67°C
For Use With (Equipment)Forma FDE, Revco RDE, HERAfreeze HDE, Thermo Scientific TDE Series (400 box capacity and larger)
Voltage230 V
HoldsBoxes (300)
Unit SizeEA