Check Order Status
Xylenes (Histological), Fisher Chemical™
Element Search
Structure Search
Xylenes (Histological), Fisher Chemical™
Have Questions?
Change view
Quantity:
4 L
1 gal.
Packaging:
View all
Aluminum Can
Catalog number
FCMX3S-4
Price (MYR)
-
Quantity:
4 L
Packaging:
Aluminum Can
Request bulk or custom format
Specifications
Chemical Name or Material
Xylenes
Cleanup Symbol
Solvent
Melting Point
-34°C
CAS
1330-20-7
ChemAlert Storage Symbol
Red
View more