Water, UHPLC-MS
Water, UHPLC-MS
Actual product may vary
Water, UHPLC-MS
Water, UHPLC-MS
Water, UHPLC-MS
Fisher Chemical
Water, UHPLC-MS
Unique signal-to-noise specification for the chemical industry where the solvent quality is linked directly to the sensitivity of the mass spectrometer. | CAS: 7732-18-5 | H2O | 18.015 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FCMW8-11 LClear Borosilicate Glass Bottle
Catalog number FCMW8-1
Price (MYR)
669.00
EA
Add to cart
Quantity:
1 L
Packaging:
Clear Borosilicate Glass Bottle
Price (MYR)
669.00
EA
Add to cart
Water, H2O, CAS Number-7732-18-5, dihydrogen oxide, dihydrogen monoxide, 1L, <-0.01 AU at 230nm, <-0.01 AU at 210nm, <-0.01 AU at 220nm, <-0.01 AU at 240nm, <-0.005 AU at 280nm, <-0.005 AU at 260nm, <-0.005 AU at 254nm, 2ppb max., 100 deg.C, 5ppb max., CHEBI:15377, Colorless, 1.000g/cm3, UHPLC-MS
RUO – Research Use Only
Specifications
Absorbance≤0.005 AU at 254nm, ≤0.005 AU at 260nm, ≤0.005 AU at 280nm, ≤0.01 AU at 210nm, ≤0.01 AU at 220nm, ≤0.01 AU at 230nm, ≤0.01 AU at 240nm
Chemical Name or MaterialWater
pH7
CAS7732-18-5
ColorColorless
Melting Point0°C
Boiling Point100°C
Recommended StorageRoom temperature
Molecular FormulaH2O
Synonymdihydrogen oxide, dihydrogen monoxide
InChI KeyXLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILESO
IUPAC Nameoxidane
Molecular Weight (g/mol)18.015
PubChem CID962
ChEBICHEBI:15377
GradeUHPLC-MS
PackagingClear Borosilicate Glass Bottle
Residue after Evaporation≤1ppm
Vapor Pressure17.5mmHg at 20°C
Density1.000 g/cm3
Quantity1 L
Physical FormLiquid
Unit SizeEA