Check Order Status
Tetrahydrofuran (Certified), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Tetrahydrofuran (Certified), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMT397-1
1 L
Glass Bottle
Catalog number
FCMT397-1
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Tetrahydrofuran
pH
7 to 8
Cleanup Symbol
Solvent
Melting Point
-108.4°C
CAS
109-99-9
View more