Check Order Status
Potassium Hydroxide Solution, 0.1N (In Isopropanol/Certified), Fisher Chemical
Element Search
Structure Search
Fisher Chemical
Potassium Hydroxide Solution, 0.1N (In Isopropanol/Certified), Fisher Chemical
For use as a titration standard
Have Questions?
Catalog Number
Quantity
Packaging
FCMST110-500
500 mL
HDPE Bottle
Catalog number
FCMST110-500
Price (MYR)
571.00
EA
Add to cart
Quantity:
500 mL
Packaging:
HDPE Bottle
Request bulk or custom format
Price (MYR)
571.00
EA
Add to cart
Specifications
Chemical Name or Material
Potassium Hydroxide Solution
Cleanup Symbol
Caustic
Name Note
0.1N, in isopropanol
Boiling Point
82.8°C
CAS
1310-58-3
View more