Check Order Status
Sodium Hydroxide Solution (0.05N (N/20)/Certified), Fisher Chemical
Element Search
Structure Search
Sodium Hydroxide Solution (0.05N (N/20)/Certified), Fisher Chemical
A volumetric standard solution, titrated at 25°C to pH 8.6 (phenolphthalein endpoint) against NIST™ potassium acid phthalate
Have Questions?
Catalog Number
Quantity
Packaging
FCMSS278-1
1 L
Poly Bottle
Catalog number
FCMSS278-1
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Sodium Hydroxide Solution
Cleanup Symbol
Caustic
Name Note
0.05N (N/20)
CAS
1310-73-2
ChemAlert Storage Symbol