Check Order Status
Sodium Hydroxide Solution (10N/Certified), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Sodium Hydroxide Solution (10N/Certified), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMSS255-1
1 L
Poly Bottle
Catalog number
FCMSS255-1
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Sodium Hydroxide Solution
pH
12.0
Cleanup Symbol
Caustic
Name Note
10N
Boiling Point
>100°C
View more