Phenolphthalein Solution, Alcoholic, 1.0%, Fisher Chemical™
Phenolphthalein Solution, Alcoholic, 1.0%, Fisher Chemical™
Phenolphthalein Solution, Alcoholic, 1.0%, Fisher Chemical™
Phenolphthalein Solution, Alcoholic, 1.0%, Fisher Chemical™
Fisher Chemical
Phenolphthalein Solution, Alcoholic, 1.0%, Fisher Chemical™
Volumetric indicator
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Packaging:
Glass Bottle
Catalog number FCMSP62-500
Price (MYR)
487.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
487.00
EA
Add to cart
Specifications
Chemical Name or MaterialPhenolphthalein Solution, Alcoholic 1.0%
Melting Point-89°C
Boiling Point83°C
CAS77-09-8
ChemAlert Storage SymbolRed
View more