Check Order Status
p-Naphtholbenzein Indicator Solution, Fisher Chemical
Element Search
Structure Search
p-Naphtholbenzein Indicator Solution, Fisher Chemical
Have Questions?
Catalog Number
Quantity
FCMSN1-500
500 mL
Catalog number
FCMSN1-500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or Material
p-Naphtholbenzein Indicator Solution
CAS
145-50-6
ChemAlert Storage Symbol
Red
CAS
108-88-3
CAS
67-63-0
View more