Check Order Status
Cleaning Solution (Chromic-Sulfuric Acid)
Actual product may vary
Cleaning Solution (Chromic-Sulfuric Acid)
Removes nearly impossible stains from glassware
Have Questions?
Catalog Number
Volume (Metric)
FCMSC88-500
500 mL
Catalog number
FCMSC88-500
Price (MYR)
524.00
EA
Add to cart
Volume (Metric):
500 mL
Price (MYR)
524.00
EA
Add to cart
Premixed
Specifications
Form
Liquid
Appearance
Dark Brown
Boiling Point
290°C
Composition
Chromium Trioxide 0.5%, Sulfuric Acid 93.87%, Water 5.63%
Volume (Metric)
500 mL