Check Order Status
Buffer Solution, pH 5.00 (Certified), Fisher Chemical™
Element Search
Structure Search
Buffer Solution, pH 5.00 (Certified), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FCMSB102-1
1 L
Catalog number
FCMSB102-1
Price (MYR)
438.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
438.00
EA
Add to cart
Chemical Identifiers
CAS
877-24-7, 1310-73-2, 7732-18-5
Specifications
Chemical Name or Material
Buffer Solution, pH 5.00
CAS
877-24-7
ChemAlert Storage Symbol
Gray
Melting Point
0°C
Boiling Point
100°C
View more