Check Order Status
Hydrochloric Acid Solution, 0.5N (N/2) (Certified), Fisher Chemical™
Element Search
Structure Search
Hydrochloric Acid Solution, 0.5N (N/2) (Certified), Fisher Chemical™
Standardized at 25°C to pH 8.2 against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate
Have Questions?
Catalog Number
Quantity
Packaging
FCMSA50-1
1 L
Poly Bottle
Catalog number
FCMSA50-1
Price (MYR)
378.00
EA
Add to cart
Quantity:
1 L
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
378.00
EA
Add to cart
Specifications
Chemical Name or Material
Hydrochloric Acid Solution, 0.5N (N/2)
pH
0.10
Cleanup Symbol
Acid
CAS
7647-01-0
ChemAlert Storage Symbol