Check Order Status
Hydrochloric Acid Concentrate, 10N ACS (Certified), Fisher Chemical
Element Search
Structure Search
Fisher Chemical
Hydrochloric Acid Concentrate, 10N ACS (Certified), Fisher Chemical
Each bottle contains exact amount of 10N concentrate required to make 1 liter of 1N or 10 liters of 0.1N (N/10) solution
Have Questions?
Catalog Number
Quantity
Packaging
FCMSA49
100 mL
Glass Bottle
Catalog number
FCMSA49
Price (MYR)
429.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
429.00
EA
Add to cart
Specifications
Chemical Name or Material
Hydrochloric Acid Concentrate, 10N
pH
1
Cleanup Symbol
Acid
Color
Colorless
CAS
7647-01-0
View more