Check Order Status
Trichloroacetic Acid Solution (20% w/v), Fisher Chemical
Element Search
Structure Search
Trichloroacetic Acid Solution (20% w/v), Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FCMSA254-500
500 mL
Glass Bottle
Catalog number
FCMSA254-500
Price (MYR)
-
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Trichloroacetic Acid Solution
Name Note
20% (w/v)
CAS
76-03-9
ChemAlert Storage Symbol
White
Melting Point
54°C
View more